Ethyl isothiocyanatoformate 95%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:16182-04-0
:
Formula:C₄H₅NO₂S
MDL Number:MFCD00004814
:131.16 g/mol
Boiling Pt:56 °C (18 mmHg)
Density:1.16 g/cm³
Flash Pt:50 °C
UN:2929
ADR:6.1,II
:GHS02
:GHS05
:GHS06
Storage Temperature:Refrigerator
Specification Test Results
- Appearance:Clear colorless to yellow to orange Liquid
- Infrared spectrum:Conforms
- Refractive index:1.4980 to 1.5020 (20°C, 589 nm)
- GC:≥94 %