Methyl-3-hydroxy-2,2-dimethylpropanoate 98+%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:14002-80-3
:
Formula:C₆H₁₂O₃
MDL Number:MFCD00009707
:132.1600 g/mol
Boiling Pt:177…178 °C (760 mmHg)
Density:1.04 g/cm³
Flash Pt:76 °C
:GHS07
Storage Temperature:Ambient
Specification Test Results
- Appearance:Clear colorless Liquid
- Infrared spectrum:Conforms
- Refractive index:1.4280 to 1.4310 (20°C, 589 nm)
- GC:≥98.0 %