3-Furanmethanol 99%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:4412-91-3
:
Formula:C₅H₆O₂
MDL Number:MFCD00005352
:98.10 g/mol
Boiling Pt:79…80 °C (17 mmHg)
Density:1.139 g/cm³
Flash Pt:38 °C
Storage Temperature:Ambient
UN:1987
ADR:3,III
:GHS07
:GHS02
Specification Test Results
- Appearance:Clear colorless to light yellow Liquid
- Infrared spectrum:Conforms
- Refractive index:1.4835 to 1.4855 (20°C, 589 nm)
- GC:≥98.5 %