

4,4,5,5-Tetramethyl-1,3,2-dioxaborolane 97% stabilised
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:25015-63-8
:
Formula:C₆H₁₃BO₂
MDL Number:MFCD00674030
:127.98 g/mol
Boiling Pt:42…43 °C (50 mmHg)
Density:0.882 g/cm³
Flash Pt:5 °C
Storage Temperature:Refrigerator
UN:3399
ADR:4.3,II
:GHS02
:GHS07
Specification Test Results
- Appearance:Clear colorless to light yellow Liquid
- Infrared spectrum:Conforms
- Refractive index:1.3950 to 1.4000 (20°C, 589 nm)
- GC:≥96.0 %
- Stabilizer:≤1.1 % Triethylamine