
Dichloro(1,5-cyclooctadiene)platinum(II) 98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:12080-32-9
:
Formula:C₈H₁₂Cl₂Pt
MDL Number:MFCD00012413
:374.17 g/mol
Melting Pt:285 °C (dec.)
:GHS07
Storage Temperature:Ambient
Specification Test Results
- Appearance:White to light yellow Crystalline powder or crystals
- Infrared spectrum:Conforms
- Assay:≥97.5 % (ex Pt)