

Kieselguhr (Celite®), Celite®, hohe Reinheit, analytische Qualität zur Analyse analytical grade
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:61790-53-2
:7631-86-9
Formula:SiO₂
MDL Number:MFCD00011232
:60.08 g/mol
:GHS07
:GHS08
Storage Temperature:Raumtemperatur
Specification Test Results
- Appearance:White to grey-pink Powder
- Loss on drying:≤0.5 %
- Additional info:typical chemical analysis :
- pH:5 to 8 (25°C, 10% aqueous suspension)
- Arsenic (As):≤10 ppm
- Lead (Pb):≤10 ppm
- Loss on ignition:≤2 %
- Conductivity:≤15 µS/cm
Specification Test Results
Frequently Bought Together