Order Entry
Germany
ContactUsLinkComponent
2-N-BOC-amino-3-(4-tetrahydropyranyl)propionic acid 95%
Irritant
2-N-BOC-amino-3-(4-tetrahydropyranyl)propionic acid 95%
Catalog # ACRO360485000
CAS Number:  
undefined
Irritant
2-N-BOC-amino-3-(4-tetrahydropyranyl)propionic acid 95%
Catalog # ACRO360485000
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    500 mg
  • Seq:
    2725

Specifications

Specification Test Results

  • Appearance:
    White to off-white Chunks
  • Infrared spectrum:
    Conforms
  • NMR:
    ≥94 %

Specification Test Results

About this item

CAS Number:182287-51-0
 :
Formula:C₁₃H₂₃NO₅
MDL Number:MFCD02683139
 :273.329 g/mol
 :GHS07
Storage Temperature:Ambient