tert-Butoxybis(dimethylamino)methane
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:5815-08-7
:
Formula:C₉H₂₂N₂O
MDL Number:MFCD00042858
:174.288 g/mol
Boiling Pt:50…55 °C (15 mmHg)
Density:0.844 g/cm³
Flash Pt:41 °C
Storage Temperature:Freezer
UN:1993
ADR:3,III
:GHS07
:GHS02
Specification Test Results
- Appearance:Clear light yellow Liquid
- Infrared spectrum:Conforms
- GC:90 % (sum of 3 constituents) varying proportionally Tris(dimethylamino)methane, N,N-dimethylformamide-N,N-di-tert.-butylacetal, tert.-Butoxybis(dimethylamino)methane