
1,5-Cyclooctadienebis(methyldiphenylphosphine)iridium(I) hexafluorophosphate 97%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:38465-86-0
:
Formula:C₃₄H₃₈F₆IrP₃
MDL Number:MFCD00064800
:845.79 g/mol
Melting Pt:224 °C
Storage Temperature:Ambient
ADR:6.1,III
:GHS07
Specification Test Results
- Appearance:Red Crystalline powder
- Infrared spectrum:Conforms
- Assay:≥96.0 %