Order Entry
Germany
ContactUsLinkComponent
 
 
2-(4-Aminophenyl)-1,1,1,3,3,3-hexafluoro-2-propanol 97%
 
undefined
 
2-(4-Aminophenyl)-1,1,1,3,3,3-hexafluoro-2-propanol 97%

 

  • Appearance:
    Light brown Crystalline powder
  • Infrared spectrum:
    Conforms
  • Melting point:
    147 to 152 °C
  • Titration with HClO4:
    ≥96.0 %