
tert-Butyl-N-[4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]carbamate 97+%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:330793-01-6
:
Formula:C₁₇H₂₆BNO₄
MDL Number:MFCD02179439
:319.208 g/mol
Melting Pt:166…169 °C
:GHS07
Storage Temperature:Refrigerator
Specification Test Results
- Appearance:White to off-white Crystalline powder
- Infrared spectrum:Conforms
- Melting point:168 to 173 °C
- HPLC:≥97.0 %