Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Germany
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Pentasodium triphosphate
penta-Sodium triphosphate 85% tech., pure
penta-Sodium triphosphate 85% tech., pure
:
Thermo Fisher Scientific
penta-Sodium triphosphate 85% tech., pure
:
Thermo Fisher Scientific
:
7758-29-4
:
:
Na₅O₁₀P₃
:
MFCD00003514
:
367.86 g/mol
:
622 °C
:
2,52 g/cm³ (20 °C)
:
GHS07
:
Ambient
Appearance:
White Crystalline powder
Titration with HClO4:
≥80 %
ACRO21867