Thioflavin T
Supplier: Thermo Fisher Scientific
About this item
CAS No.: 2390-54-7
Synonyms: CI 49005, Thioflavine T, Basic Yellow 1, 2-(4-(Dimethylamino)phenyl)-3,6-dimethylbenzo[d]thiazol-3-ium chloride, Setoflavine T
- CAS Number:2390-54-7
- Formula:C₁₇H₁₉ClN₂S
- MDL Number:MFCD00011944
- Molecular Weight:318.86 g/mol
- Storage Temperature:Ambient
3 Options Available Below
Product Details & Documents
CAS No.: 2390-54-7
Synonyms: CI 49005, Thioflavine T, Basic Yellow 1, 2-(4-(Dimethylamino)phenyl)-3,6-dimethylbenzo[d]thiazol-3-ium chloride, Setoflavine T
- CAS Number:2390-54-7
- Formula:C₁₇H₁₉ClN₂S
- MDL Number:MFCD00011944
- Molecular Weight:318.86 g/mol
- Storage Temperature:Ambient
Documents
ACRO21176
SDS
Specifications for Product Family
Specification Test Results
- Appearance:Yellow to golden to orange-brown Powder
- Infrared spectrum:Conforms
- Solubility:(1 mg/ml in methanol) Clear to slightly hazy yellow to yellow-green
- UV Lambda max:409 to 415 nm (10 mg/l in water)
Product Family Options
Product Information
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



