Remazol brilliant blue R, pure
ACRO207550250
:
- Pack type:Glass bottle
- Pk:25 g
- Seq:3308
- Appearance:Dark blue Crystalline powder
- Infrared spectrum:Conforms
- UV spectrum:Complies
:
:626.533 g/mol
:Ambient
:2580-78-1
:C₂₂H₁₆N₂Na₂O₁₁S₃
:MFCD00001215