ContactUsLinkComponent
Poly(methyl methacrylate), M.W. ∼15000 (GPC)
Poly(methyl methacrylate), M.W. ∼15000 (GPC)
Certificates
undefined
Poly(methyl methacrylate), M.W. ∼15000 (GPC)
Specification Test Results
  • Appearance:
    White Beads
  • Infrared spectrum:
    Conforms
  • Loss on drying:
    ≤3 %
  • Additional info:
    bead size (typically): 10 to 100 micron