
Diethyl-bis(hydroxymethyl)malonate 95%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:20605-01-0
:
Formula:C₉H₁₆O₆
MDL Number:MFCD00009130
:220.221 g/mol
Melting Pt:45…52 °C
Flash Pt:>110 °C
:GHS07
Storage Temperature:Ambient
Specification Test Results
- Appearance:White to off-white Crystalline solid
- Infrared spectrum:Conforms
- Melting point:45 to 52 °C
- GC:≥94 %
- Titration with HClO4:97.0 to 102.0 %