β-D-Ribofuranose-1,2,3,5-tetraacetate 98+%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:13035-61-5
:
Formula:C₁₃H₁₈O₉
MDL Number:MFCD00005358
:318.28 g/mol
Melting Pt:79…81 °C
Storage Temperature:Ambient
Specification Test Results
- Appearance:White to almost white Crystalline powder
- Infrared spectrum:Conforms
- Loss on drying:≤1 % (60°C, 2 h) (vacuum)
- HPLC:≥98.0 %
- Residue after ignition:≤0.1 %
- Specific optical rotation:-12° to -14° (20°C, 589 nm) (c=10, CHCl3)
- Heavy metals (as Pb):≤20 ppm