
1-Benzylpiperidin-3-one hydrochloride hydrate, hydrate 95%
Catalog # ACRO166920010
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:1 g
- Seq:3047
Specifications
Specification Test Results
- Appearance:Cream to light brown Powder
- Infrared spectrum:Conforms
- Titration Argentometric:98 to 102 % (on dry substance)
- Water:≤8 % (K.F.)
- GC:≥94 % (on free base)
Specification Test Results
About this item
:
:225.72 g/mol (anhy)
Melting Pt:170…176 °C (dec.)
Storage Temperature:Ambient
UN:2811
ADR:6.1,III
:GHS07
CAS Number:50606-58-1
Formula:C₁₂H₁₅NO.HCl.xH₂O
MDL Number:MFCD00150096