Digital Solutions
Applications
Support
About
Germany
ContactUsLinkComponent
Silicone oil
Silicone oil, 475 -525 mm²/s (25°C) for melting point and boiling point apparatuses
Silicone oil, 475 -525 mm²/s (25°C) for melting point and boiling point apparatuses
:
Thermo Fisher Scientific
Silicone oil, 475 -525 mm²/s (25°C) for melting point and boiling point apparatuses
:
Thermo Fisher Scientific
:
68083-14-7
:
63148-62-9
:
MFCD00132673
:
–50 °C
:
0.96
:
300 °C
:
Ambient
ACRO16385