Order Entry
Germany
ContactUsLinkComponent
 
 
Ethyl-1-benzyl-3-oxopiperidine-4-carboxylate hydrochloride 97% tech.
 
undefined
 
Ethyl-1-benzyl-3-oxopiperidine-4-carboxylate hydrochloride 97% tech.

 

  • Appearance:
    White to brown Crystalline powder
  • Infrared spectrum:
    Conforms
  • Melting point:
    160 to 165 °C
  • Titration with HClO4:
    ≥96.0 %