Order Entry
Germany
ContactUsLinkComponent
Scopolamine hydrobromide trihydrate 99%
Toxic
Scopolamine hydrobromide trihydrate 99%
Certificates
undefined
Toxic
Scopolamine hydrobromide trihydrate 99%

Specification Test Results

  • Appearance:
    Colorless to white Crystalline powder or crystals
  • Infrared spectrum:
    Conforms
  • Water:
    10 to 13 % (K.F.)
  • Specific optical rotation:
    -24° to -27° (20°C, 589 nm) (c=5, H2O) (on anhydrous substance)
  • Titration with HClO4:
    ≥98.5 %

Specification Test Results

Related Information

Frequently Bought Together