5-Methoxy-2-methyl-3-indoleacetic acid 98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:2882-15-7
:
Formula:C₁₂H₁₃NO₃
MDL Number:MFCD00005618
:219.24 g/mol
Melting Pt:162…165 °C
Storage Temperature:Refrigerator
Specification Test Results
- Appearance:Off-white to light yellow to light beige Crystalline powder
- Infrared spectrum:Conforms
- Melting point:161 to 166 °C
- Titration with NaOH:≥97.5 %
- Elemental Analysis:12.00 to 12.62 % N
- Specific optical rotation:+52° to +54° (20 °C, 589 nm) (c=1 in DMF)