Yohimbine hydrochloride 99%
:65-19-0
:
:C₂₁H₂₆N₂O₃
:MFCD00012674
:390.91 g/mol
:285…288 °C (dec.)
:2811
:6.1,II
:GHS06
:GHS08
:Ambient
- Appearance:White to light yellow Powder
- Infrared spectrum:Conforms
- Specific optical rotation:+99° to +105° (20°C, 589 nm) (c=1, H2O)
- Titration with HClO4:≥98.5 %