Nɑ-Tosyl-L-arginine methyl ester hydrochloride 98+%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:1784-03-8
:
Formula:C₁₄H₂₂N₄O₄S·HCl
MDL Number:MFCD00012578
:378.87 g/mol
Melting Pt:146…149 °C
Storage Temperature:Ambient
Specification Test Results
- Appearance:White Crystalline powder
- Infrared spectrum:Conforms
- Melting point:146 to 149 °C
- Titration with NaOH:98.0 to 102.0 %