Order Entry
Germany
ContactUsLinkComponent
 
Palladium 10% on activated carbon wetted with ca. 50% water, unreduced
 
undefined
Palladium 10% on activated carbon wetted with ca. 50% water, unreduced

 

  • Appearance:
    Black Powder
  • Assay:
    9.7 to 10.3 % (Pd)
  • Loss on drying:
    45 % to 55 %
  • GC:
    ≥97.5 %

 

 

Frequently Bought Together