Rhodamine 110 98+%, pure laser grade
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:13558-31-1
:
Formula:C₂₀H₁₅ClN₂O₃
MDL Number:MFCD00042009
:366.801 g/mol
Melting Pt:>360 °C
Storage Temperature:Ambient
Specification Test Results
- Appearance:Green to red to red-brown Crystalline powder or crystals
- Infrared spectrum:Conforms
- HPLC:≥98.0 %
- UV Lambda max:498 to 504 nm (c=10 mg/l in Methanol)
- Suitability:(for use as a laser dye) Passes test
- Molar absorption coefficient:≥77000 l/(mol.cm) (at Lambda max)