Order Entry
Germany
ContactUsLinkComponent
Petrolatum, white, pure
Petrolatum, white, pure
Certificates
undefined
Petrolatum, white, pure
Certificates

Specification Test Results

  • Appearance:
    White Paste
  • Infrared spectrum:
    Conforms
  • Viscosity:
    5 to 8 mm²/s (at 100°C)
  • Solidification point:
    50 to 56°C

Specification Test Results

Related Information

Certificates
Search by Lot Number