
Pyranine 98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:6358-69-6
:
Formula:C₁₆H₇Na₃O₁₀S₃
MDL Number:MFCD00037575
Density:2.15 g/cm³
:524.371 g/mol
:GHS07
Storage Temperature:Ambient
Specification Test Results
- Appearance:Yellow to yellow-green Powder or crystals
- Infrared spectrum:Conforms
- UV spectrophotometry:≥97.5 %
- UV Lambda max:450 to 458 nm (in 0.01N NaOH)