Order Entry
Germany
ContactUsLinkComponent
2-Hydroxypropyl methacrylate (mixture of isomers) 97+% stabilised
Irritant
2-Hydroxypropyl methacrylate (mixture of isomers) 97+% stabilised
Certificates
undefined
Irritant
2-Hydroxypropyl methacrylate (mixture of isomers) 97+% stabilised

Specification Test Results

  • Appearance:
    Clear liquid
  • Infrared spectrum:
    Conforms
  • Refractive index:
    1.4460 to 1.4490 (20°C, 589 nm)
  • GC:
    ≥97.0 % (Mixture of isomers)
  • Color scale:
    ≤20 APHA
  • Stabilizer:
    150 to 300 ppm MEHQ

Specification Test Results

Related Information