
1,1'-Diethyl-2,2'-cyanine Iodide 99%
Catalog # ACRO407255000
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:500 mg
- Seq:2725
Specifications
Specification Test Results
- Appearance:Golden to dark red Crystalline powder
- Infrared spectrum:Conforms
- UV spectrophotometry:≥98.5 %
Specification Test Results
About this item
:
:454.36 g/mol
Melting Pt:273 °C
Storage Temperature:Ambient
UN:2811
ADR:6.1,II
:GHS06
CAS Number:977-96-8
Formula:C₂₃H₂₃IN₂
MDL Number:MFCD00011971