Order Entry
Germany
ContactUsLinkComponent
 
 
5-Chlorothiophene-2-carboxaldehyde 97%
 
undefined
 
5-Chlorothiophene-2-carboxaldehyde 97%

 

  • Appearance:
    Clear yellow to orange-red Liquid (May darken on storage)
  • Infrared spectrum:
    Conforms
  • Refractive index:
    1.6040 to 1.6070 (20°C, 589 nm)
  • GC:
    ≥96.0 %