Order Entry
Germany
Orders LinkContactUsLinkComponent
 

Ethylene glycol monostearate

Ethylene glycol monostearate
 
Ethylene glycol monostearate
Ethylene glycol monostearate

 

 

Appearance: White to light yellow Flakes

 :  2-Hydroxyethyl stearate
  •  :111-60-4
  •  :C₂₀H₄₀O₃
  •  :MFCD00051465
  •  :328.5298 g/mol
  •  :149 °C
  •  :55…60 °C
  •  :Ambient

 

 

 

  • Appearance:
    White to light yellow Flakes
  • Infrared spectrum:
    Conforms
  • Melting point:
    55 to 60 °C
  • Additional info:
    compound is a mixture of various chain-length esters and di(ethylene glycol) ester
Product Family Options

 

  • Pack type
    Pk
     
     
  • Glass bottle
    100 g
       
       
    1 * 100 g
    1 * 100 g - 31,40 €
    31,40 €
    1 * 100 g
       31,40 €

  • Glass bottle
    250 g
       
       
    1 * 250 g
    1 * 250 g - 56,80 €
    56,80 €
    1 * 250 g
       56,80 €

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.