
N,N'-Bis(2-hydroxyethyl)ethylenediamine 95%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:4439-20-7
:
Formula:C₆H₁₆N₂O₂
MDL Number:MFCD00002842
:148.21 g/mol
Melting Pt:92…99 °C
Storage Temperature:Ambient
UN:3259
ADR:8,III
:GHS05
Specification Test Results
- Appearance:White to light yellow to beige Powder or crystals and/or chunks
- Infrared spectrum:Conforms
- Melting point:94 to 101 °C
- Titration with HClO4:94 to 106 %