Order Entry
Germany
ContactUsLinkComponent
3-[(4S)-2,2-Dimethyl-1,3-dioxolan-4-yl]propanol 97%
Irritant
3-[(4S)-2,2-Dimethyl-1,3-dioxolan-4-yl]propanol 97%
Catalog # ACRO339610010
CAS Number:  
undefined
Irritant
3-[(4S)-2,2-Dimethyl-1,3-dioxolan-4-yl]propanol 97%
Catalog # ACRO339610010
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

Specification Test Results

  • Appearance:
    Clear colorless to yellow Viscous liquid
  • Infrared spectrum:
    Conforms
  • Refractive index:
    1.4410 to 1.4430 (20°C, 589 nm)
  • GC:
    ≥96.0 %
  • Specific optical rotation:
    +9° to +12° (20°C, 589 nm) (c=1, CHCl3)

Specification Test Results

About this item

 :
 :160.213 g/mol
Boiling Pt:90 °C (10 mmHg)
 :GHS07
Storage Temperature:Ambient
CAS Number:51268-87-2
Formula:C₈H₁₆O₃
MDL Number:MFCD01631673