Order Entry
Germany
ContactUsLinkComponent
Dimethyloctadecyl[3-(trimethoxysilyl)propyl]ammonium chloride 60% in methanol
Flammable
Toxic
Health
Dimethyloctadecyl[3-(trimethoxysilyl)propyl]ammonium chloride 60% in methanol
Catalog # ACRO338531000
CAS Number:  
undefined
Flammable
Toxic
Health
Dimethyloctadecyl[3-(trimethoxysilyl)propyl]ammonium chloride 60% in methanol
Catalog # ACRO338531000
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    100 mL
  • Seq:
    2440

Specifications

Specification Test Results

  • Appearance:
    Clear yellow Solution
  • Infrared spectrum:
    Conforms
  • Titration Argentometric:
    ≥58 %

Specification Test Results

About this item

 :
 :496.289 g/mol
Density:0.89 g/cm³
Flash Pt:15 °C
Storage Temperature:Ambient
UN:1230
ADR:3,II
 :GHS02
 :GHS06
 :GHS08
CAS Number:27668-52-6
Formula:C₂₆H₅₈ClNO₃Si
MDL Number:MFCD00054224