N-Boc-DL-2-aminotetralin-2-carboxylic acid 98%
Catalog # ACRO334370010
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:1 g
- Seq:3047
Specifications
Specification Test Results
- Appearance:White to beige Crystalline powder
- Infrared spectrum:Conforms
- HPLC:≥97.5 %
Specification Test Results
About this item
:
:291.347 g/mol
Melting Pt:177…187 °C
:GHS07
Storage Temperature:Ambient
CAS Number:98569-12-1
Formula:C₁₆H₂₁NO₄
MDL Number:MFCD00800586