
(E)-4-(Dimethylamino)but-3-en-2-one 98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:2802-08-6
:
Formula:C₆H₁₁NO
MDL Number:MFCD00142611
:113.16 g/mol
Boiling Pt:101…102 °C (5 mmHg)
Density:0.96 g/cm³
Flash Pt:107 °C
:GHS07
Storage Temperature:Ambient
Specification Test Results
- Appearance:Clear red-brown to dark brown Liquid
- Infrared spectrum:Conforms
- Refractive index:1.5570 to 1.5600 (20°C, 589 nm)
- GC:≥97.5 %
- Titration with HClO4:≥97.5 %