2-Ethynylpyridine 98+%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:1945-84-2
:
Formula:C₇H₅N
MDL Number:MFCD00041598
:103.12 g/mol
Boiling Pt:77…78 °C (14 mmHg)
Density:1.02 g/cm³
Flash Pt:76 °C
Storage Temperature:Refrigerator
Specification Test Results
- Appearance:Clear dark brown Liquid
- Infrared spectrum:Conforms
- GC:≥98.0 %