Carbopol® 940
278380000.
:
- Pack type:Plastic bottle
- Pk:1
- Seq:0001
- Appearance:White Powder
- Infrared spectrum:Conforms
- Loss on drying:≤2 %
- Heavy metals:≤10 ppm (Hg, Pb, As, Sb)
- Viscosity:40000 to 60000 cP (at 25°C) (Brookfield, 20 rpm) (neutralized soln., 0.5% dispersion)
- Clarity of solution:≥85 % transmission (neutralized soln., 420 nm, 0.5% dispersion)
- Residual solvents:≤0.5 % Benzene
:
:Ambient
:9003-01-4
:MFCD01941675