Quinoline yellow sulphonic acid sodium salt (Acid yellow 3) 95%, water soluble, pure
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:8004-92-0
:
Formula:C₁₈H₁₁NO₂.2NaO₃S
MDL Number:MFCD00080727
:GHS07
Storage Temperature:Ambient
Specification Test Results
- Appearance:Yellow to orange-brown Powder
- Infrared spectrum:Conforms
- HPLC:≥94 %