Order Entry
Germany
ContactUsLinkComponent
 
3-Methyl-5-methoxycarbonyl-4-piperidone hydrochloride 99%
Catalog # ACRO268085000
CAS Number:  
undefined
3-Methyl-5-methoxycarbonyl-4-piperidone hydrochloride 99%
Catalog # ACRO268085000
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    500 mg
  • Seq:
    2725

Specifications

Specification Test Results

  • Appearance:
    White Crystalline powder
  • Infrared spectrum:
    Conforms
  • Titration Argentometric:
    ≥98.5 %
  • Titration with HClO4:
    ≥98.5 %

Specification Test Results

About this item

 :
Melting Pt:160…175 °C
 :207.654 g/mol
Storage Temperature:Ambient
CAS Number:205692-64-4
Formula:C₈H₁₄ClNO₃
MDL Number:MFCD00143176