Goat anti-human IgA reacts specifically with IgA heavy chains (α chains) and not with immunoglobulin light chains. To minimize crossreactivity, it has been adsorbed human IgG prior to conjugation. This conjugate is labeled with CF™633 dye. CF™ dyes offer advantages in brightness, photostability, and specificity compared to other fluorescent dyes. Far-red fluorescent CF™633 (Ex/Em 630/650 nm) produces brighter antibody conjugates than Alexa Fluor® 647, and has unmatched photostability.
Alexa Fluor® is a registered trademark of Thermo Fisher Scientific. Cy®5 is a registered trademark of GE Healthcare.
Alexa Fluor® and Texas Red® are registered trademarks of Thermo Fisher Scientific.
Alexa Fluor® is a registered trademark of Thermo Fisher Scientific.
Type: Secondary
Antigen: IgA
Clonality: Polyclonal
Clone:
Conjugation: CF633
Epitope:
Host: Goat
Isotype:
Reactivity: Human
- Catalog No:
- BTIU20427-50UL
- BTIU20427-500UL
- Antigen symbol:
- IgA
- IgA
- Antigen name:
- Human IgA
- Human IgA
- Antibody type:
- Secondary
- Secondary
- Clonality:
- Polyclonal
- Polyclonal
- Conjugation:
- CF® 633
- CF® 633
- Reactivity:
- Human
- Human
- Host:
- Goat
- Goat
- Form:
- Liquid
- Liquid
- Purification:
- Affinity purified
- Affinity purified
- Concentration:
- 2 mg/mL
- 2 mg/mL
- Cross adsorption:
- Human IgG
- Human IgG
- Storage buffer:
- PBS, 50% glycerol, 2 mg/ml BSA, 0,05% sodium azide
- PBS, 50% glycerol, 2 mg/ml BSA, 0,05% sodium azide
- Storage temperature:
- Store at –20 °C.
- Store at –20 °C.