Order Entry
Germany
ContactUsLinkComponent
Anti-Calcitonin Antibody
Anti-Calcitonin Antibody
Catalog # BSBTPB9937
Supplier:  Boster Bio
undefined
Anti-Calcitonin Antibody
Catalog # BSBTPB9937
Supplier:  Boster Bio

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    calcitonin/calcitonin-related polypeptide, alpha
  • Antigen symbol:
    CALCA
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    Rat,
    Mouse
  • Cross adsorption:
    No
  • Gene ID:
    12310
  • Antigen synonyms:
    CGRP-1|Ctn|CA|CGRP1|Ct|Calc|Cgrp|Calc1
  • Size:
    100 µg
  • Pk:
    100 µG

Specifications

About this item

Rabbit IgG polyclonal antibody for Calcitonin gene-related peptide 1(CALCA) detection. Tested with IHC-P in Mouse;Rat.

Type: Primary
Antigen: CALCA
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse, Rat