Vacuum manifold, E-Z 96®
VAC-03
- Description:E-Z 96® vacuum manifold
- Pk:1
The E-Z 96® vacuum manifold is designed for DNA/RNA sample preparation with E-Z 96® DNA/RNA plates.
- Upper plate has a machined recess that fits E-Z 96® filter plate and contains a custom gasket
- Top plate has locating holes which correspond directly to the locating pins in the bottom plate for easy alignment and stability
The chamber can be used either as a simple dump for waste filtrate or as a receptacle for a standard 96-well microtitre plate (collection plate).