ContactUsLinkComponent
Immobilised GST agarose, Pierce™
Immobilised GST agarose, Pierce™
  PIER20211
 :  
undefined
Immobilised GST agarose, Pierce™
  PIER20211
 :  
  • Pk:
    5 ml
  • Description:
    Immobilised GST agarose
Immobilised GST is glutathione S-transferase from Schistosoma japonicum that has been covalently immobilised on crosslinked 6% beaded agarose.
  • Provides purification of specific antibodies from those directed against the GST tag
  • Bound antibodies can be eluted with 6 M guanidinium chloride allowing for up to10 purifications with the support
Antibody molecules recognising the GST portion of a recombinant fusion protein can be removed from the desired antibody by passing antiserum over an immobilised GST column. GST antibodies will bind to the immobilised GST column, whereas antibodies against other fusion proteins will pass through the column unrestrained.