Pierce™ OPD (o-phenylenediamine Dihydrochloride)
Supplier: Thermo Fisher Scientific
Certificates
About this item
OPD (o-phenylenediamine dihydrochloride) is a water-soluble substrate for horseradish peroxidase (HRP) that produces a yellow-orange product detectable at 492 nm by ELISA plate readers.