ContactUsLinkComponent
Affinity purification kits, Melon™ gel Monoclonal IgG purification kit
Affinity purification kits, Melon™ gel Monoclonal IgG purification kit
Catalog # PIER45214
CAS Number:  
undefined
Affinity purification kits, Melon™ gel Monoclonal IgG purification kit
Catalog # PIER45214
CAS Number:  
Specifications
  • Pk:
    1 KIT
  • Description:
    Melon™ Gel Monoclonal IgG Purification Kit, sufficient for IgG purification up to 1 l of cell culture supernatant or up to 200 ml of ascites fluid
Specifications
About this item
Melon™ Gel Monoclonal IgG Purification Kit is a high-yield, mild purification system for monoclonal antibodies from ascites or hybridoma cell culture supernatant, or for large scale antibody purification from serum.
  • Higher recovery and purity
  • Effective for subclasses of IgG that bind poorly to Protein A or G
  • Regenerable, support can be regenerated and used for up to 6 purifications
  • Compatible, amine free buffer does not require removal or neutralisation
Delivery: Kit includes Melon™ gel support, 200 ml; Melon™ gel purification buffer (dry mix to make 100X), 1 L; and Melon™ gel regenerant (dry mix to make 5X), 1 L.