Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Germany
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Ammonium iron (III) sulphate
Ammonium iron(III) sulphate USP Test Solution (TS)
Ammonium iron(III) sulphate USP Test Solution (TS)
Toxic
Corrosive
Health
Environment
Ammonium iron(III) sulphate USP Test Solution (TS)
Supplier:
VWR Chemicals
Certificates
Expand image
Toxic
Corrosive
Health
Environment
Ammonium iron(III) sulphate USP Test Solution (TS)
Supplier:
VWR Chemicals
Certificates
About this item
Merck Index:
12,00549
Related Information
Documentation
Certificates
Search by Lot Number
CoC_VWRC85346.260_230200
VWRC85346.180_250029.pdf
VWRC85346.180_250070.pdf
VWRC85346.260_250029.pdf
VWRC85346.260_250070.pdf
SDS
sds_VWRC85346
Product Literature
VWR Chemicals ACS Reag PH EUR Substances, Standards, Solutions and Reagents
Documentation