Pierce™ Antibody Purification Kits, Chicken IgY Purification Kit
Catalog # PIER44918
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Pk:1 KIT
- Description:Pierce™ chicken IgY purification kit, sufficient to purify IgY from 5 egg yolks
- Seq:01
Specifications
About this item
Pierce™ chicken IgY purification kit purifies chicken antibodies from hen egg yolks using a unique delipidation reagent to separate protein from lipid and a precipitation reagent to selectively isolate the IgY fraction.
- Purifies twice the amount of IgY and obtain 80 to 120 mg IgY per egg yolk at 85% to 95% purity
- Simple precipitation method purifies chicken IgY without affinity columns
- Eggs can be stored in buffer and purified at a later date
Delivery: Kit includes delipidation reagent (500 ml or 5×500 ml), IgY precipitation reagent (500 ml or 5×500 ml), egg separator, (1 or 5 separators).