ContactUsLinkComponent
HRP substrate, 1-Step™ ABTS Substrate Solution
HRP substrate, 1-Step™ ABTS Substrate Solution
Catalog # PIER37615
CAS Number:  
HRP substrate, 1-Step™ ABTS Substrate Solution
Catalog # PIER37615
CAS Number:  
Specifications
  • Pk:
    250 ml
  • Description:
    1-Step™ ABTS Substrate Solution
  • Seq:
    01
Specifications
About this item
The 1-Step™ ABTS (2,2'-azinobis [3-ethylbenzthiazoline-6-sulfonic acid] diammonium salt) is a ready-to-use, water-soluble ELISA substrate for horseradish peroxidase. This substrate yields a green product with absorbance maxima at 410 nm and 650 nm. Adding SDS to the substrate stops the reaction without changing the colour.